Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ILK
Entrez Gene ID
3611
Gene Name
integrin linked kinase
Gene Aliases
HEL-S-28, ILK-1, ILK-2, P59, p59ILK
Chromosome Location
Chr.11: 6603708 - 6610874 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014795.2 NP_001014795.1 1 - 1 219 87
GenBank mRNA AB209416.1 - 1 - 1 219 87
CR749220.1 - 1 - 1 213 87
DC379936.1 - 1 - 1 219 87
EU794625.1 - 1 - 1 52 87

View Full Product Details