The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001019.4 | NP_001010.2 | 3 - 4 | 316 | 165 |
NM_001030009.1 | NP_001025180.1 | 3 - 4 | 250 | 165 | |
GenBank mRNA | AB062400.1 | - | 1 - 1 | 232 | 165 |
AK130967.1 | - | 3 - 4 | 250 | 165 | |
AK291988.1 | - | 3 - 4 | 316 | 165 | |
AK311976.1 | - | 1 - 1 | 250 | 165 | |
BC001697.2 | - | 3 - 4 | 285 | 165 | |
BC030569.1 | - | 2 - 3 | 248 | 165 | |
BC046113.1 | - | 1 - 1 | 233 | 165 | |
BC105273.1 | - | 1 - 1 | 256 | 165 | |
BC105292.1 | - | 3 - 4 | 285 | 165 | |
BF678907.1 | - | 3 - 4 | 317 | 165 | |
BG035607.1 | - | - | 303 | 165 | |
X84407.1 | - | 1 - 1 | 220 | 165 |