Product Details

Assay Reference Genome
Location

Chr.1:196977330 on build GRCh38
Cytoband
1q31.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
81494
Gene Symbol
CFHR5
Gene Name
complement factor H related 5
Gene Aliases
CFHL5, CFHR5D, FHR-5, FHR5
Location
Chr.1:196975022-197009725 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CFHR5 XM_011510020.2 XP_011508322.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3891048 Chr1:196971388 - 196977645 on Build GRCh38 Loss CFHR5
dgv12e55 Chr1:196762018 - 197042447 on Build GRCh38 Gain CFHR2 CFHR3 CFHR4 CFHR1 F13B CFHR5
nsv469599 Chr1:196827346 - 196986100 on Build GRCh38 Loss CFHR2 CFHR4 CFHR1 CFHR5
nsv428290 Chr1:196621707 - 196992672 on Build GRCh38 Gain+Loss CFHR2 CFHR3 CFHR4 CFH CFHR1 CFHR5
nsv524689 Chr1:196972363 - 196977645 on Build GRCh38 Loss CFHR5
esv3588486 Chr1:196970045 - 196983172 on Build GRCh38 Loss CFHR5
nsv471614 Chr1:196700381 - 196986100 on Build GRCh38 Gain+Loss CFHR2 CFHR3 CFHR4 CFH CFHR1 CFHR5
esv2721051 Chr1:196970051 - 196983169 on Build GRCh38 Deletion CFHR5
nsv528460 Chr1:196573437 - 197477572 on Build GRCh38 Gain CRB1 CFHR2 MIR4735 CFHR3 CFHR4 CFH KCNT2 CFHR1 F13B CFHR5 ZBTB41 ASPM
nsv8702 Chr1:196974722 - 196985445 on Build GRCh38 Loss CFHR5

View Full Product Details