Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR128-1
- Entrez Gene ID
-
406915
- Gene Name
- microRNA 128-1
- Gene Aliases
-
MIR128A, MIRN128-1, MIRN128A, mir-128-1
- Chromosome Location
-
Chr.2: 135665397 - 135665478 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029672.1 |
|
1 - 1 |
38 |
74 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA063295.1 |
- |
1 |
266 |
|
Chr. 2 - 135665169 - 135665731 [+] on Build GRCh38 |
74 |
GenBankmRNA |
AU253801.1 |
- |
1 |
113 |
|
Chr. 2 - 135665322 - 135665964 [+] on Build GRCh38 |
74 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger RANBP2-type containing 3 |
ZRANB3 |
4933425L19Rik, AH2 |
-134.1kb
|
Anti-sense |
Chr. - 135164703 - 135531322 on Build GRCh38 |
Downstream |
UBX domain protein 4 |
UBXN4 |
UBXD2, UBXDC1, erasin |
76.1kb
|
Sense |
Chr. - 135741619 - 135785063 on Build GRCh38 |
View Full Product Details