The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_029706.1 | 1 - 1 | 39 | 68 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | transport and golgi organization 2 homolog | TANGO2 | C22orf25, MECRCN | Sense | Chr. - 20017000 - 20067164 on Build GRCh38 | |
Downstream | DGCR8 microprocessor complex subunit | DGCR8 | C22orf12, DGCRK6, Gy1, pasha | 47.0kb | Sense | Chr. - 20080232 - 20111877 on Build GRCh38 |