Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HSP90AA4P
- Entrez Gene ID
-
3323
- Gene Name
- heat shock protein 90 alpha family class A member 4, pseudogene
- Gene Aliases
-
HSP90Ad, HSPCAL2
- Chromosome Location
-
Chr.4: 189472906 - 189475853 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY956760.1 |
- |
2 - 3 |
609 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101930028 |
LOC101930028 |
|
-655.1kb
|
Anti-sense |
Chr. - 188768999 - 188818485 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 1262 |
LINC01262 |
TCONS_l2_00021807 |
184.9kb
|
Sense |
Chr. - 189659606 - 189661486 on Build GRCh38 |
View Full Product Details