Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORA14B
- Entrez Gene ID
-
677802
- Gene Name
- small nucleolar RNA, H/ACA box 14B
- Gene Aliases
-
ACA14b
- Chromosome Location
-
Chr.1: 235127803 - 235127937 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002956.1 |
|
1 - 1 |
68 |
102 |
GenBank mRNA |
AJ609457.1 |
- |
1 - 1 |
68 |
102 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA248657.1 |
- |
1 |
317 |
|
Chr. 1 - 235122341 - 235128186 [-] on Build GRCh38 |
102 |
GenBankmRNA |
DA188431.1 |
- |
1 |
325 |
|
Chr. 1 - 235127661 - 235128193 [-] on Build GRCh38 |
102 |
GenBankmRNA |
DA225811.1 |
- |
1 |
321 |
|
Chr. 1 - 235127656 - 235128190 [-] on Build GRCh38 |
102 |
GenBankmRNA |
DA115599.1 |
- |
1 |
157 |
|
Chr. 1 - 235113878 - 235128026 [-] on Build GRCh38 |
102 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 1348 |
LINC01348 |
|
-58.0kb
|
Sense |
Chr. - 235065474 - 235069818 on Build GRCh38 |
Downstream |
RNA binding motif protein 34 |
RBM34 |
|
3.2kb
|
Sense |
Chr. - 235131183 - 235161616 on Build GRCh38 |
View Full Product Details