Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR103A2
- Entrez Gene ID
-
406896
- Gene Name
- microRNA 103a-2
- Gene Aliases
-
MIR103-2, MIRN103-2, mir-103a-2
- Chromosome Location
-
Chr.20: 3917494 - 3917571 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029519.1 |
|
1 - 1 |
41 |
77 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
EL948846.1 |
- |
1 |
104 |
|
Chr. 20 - 3917451 - 3918013 [+] on Build GRCh38 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 103b-2 |
MIR103B2 |
MIR103-2-AS, MIR103-2AS, MIRN103-2AS, hsa-mir-103-2-as |
|
Anti-sense |
Chr. - 3917502 - 3917563 on Build GRCh38 |
Downstream |
ring finger protein 24 |
RNF24 |
G1L |
12.3kb
|
Anti-sense |
Chr. - 3927309 - 4015591 on Build GRCh38 |
View Full Product Details