The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204514.1 | NP_001191443.1 | 11 - 12 | 1619 | 90 |
NM_001204516.1 | NP_001191445.1 | 9 - 10 | 1323 | 90 | |
NM_001204517.1 | NP_001191446.1 | 10 - 11 | 2564 | 90 | |
NM_014970.3 | NP_055785.2 | 10 - 11 | 1455 | 90 | |
XM_005244970.1 | XP_005245027.1 | 10 - 11 | 1458 | 90 | |
XM_011509307.2 | XP_011507609.2 | 10 - 11 | 1458 | 90 | |
GenBank mRNA | AK026471.1 | - | 10 - 11 | 1234 | 90 |
AK122897.1 | - | 10 - 11 | 2566 | 90 | |
AK297368.1 | - | 9 - 10 | 1250 | 90 | |
AK301912.1 | - | 3 - 4 | 422 | 90 | |
AK303052.1 | - | 11 - 12 | 1576 | 90 | |
AK315586.1 | - | 10 - 11 | 1369 | 90 | |
BC028679.2 | - | 10 - 11 | 1412 | 90 | |
BC048012.1 | - | 9 - 10 | 1367 | 90 | |
U59919.1 | - | 10 - 11 | 1372 | 90 |