The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195430.1 | NP_001182359.1 | 2 - 3 | 445 | 73 |
XM_017020848.1 | XP_016876337.1 | 6 - 7 | 1269 | 73 | |
GenBank mRNA | CR627093.1 | - | 2 - 3 | 264 | 73 |