Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT3A
Entrez Gene ID
89780
Gene Name
Wnt family member 3A
Gene Aliases
-
Chromosome Location
Chr.1: 228007022 - 228067113 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033131.3 NP_149122.1 2 - 3 424 74
XM_011544319.2 XP_011542621.1 2 - 3 395 74
GenBank mRNA AB060284.1 - 2 - 3 322 74
AK056278.1 - 2 - 3 395 74
BC103921.1 - 2 - 3 364 74
BC103922.2 - 2 - 3 364 74
BC103923.1 - 2 - 3 364 74
DA742143.1 - 2 - 3 424 74

View Full Product Details