The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006610.3 | NP_006601.2 | 2 - 3 | 260 | 76 |
NM_139208.2 | NP_631947.1 | 2 - 3 | 260 | 76 | |
XM_017000097.1 | XP_016855586.1 | 2 - 3 | 260 | 76 | |
GenBank mRNA | AB008047.1 | - | 2 - 3 | 254 | 76 |
AK290823.1 | - | 2 - 3 | 260 | 76 | |
BC080556.1 | - | 2 - 3 | 248 | 76 | |
DA638135.1 | - | 2 - 3 | 260 | 76 | |
X98400.1 | - | 2 - 3 | 264 | 76 | |
Y09926.1 | - | 2 - 3 | 244 | 76 | |
Y18281.1 | - | 2 - 3 | 249 | 76 | |
Y18282.1 | - | 1 - 2 | 198 | 76 | |
Y18283.1 | - | 2 - 3 | 243 | 76 | |
Y18284.1 | - | 2 - 3 | 249 | 76 |