The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020435.3 | NP_065168.2 | 2 - 2 | 343 | 102 |
GenBank mRNA | BC035840.1 | - | 2 - 2 | 97 | 102 |
BC089439.1 | - | 2 - 2 | 343 | 102 | |
CF132637.1 | - | 2 - 2 | 205 | 102 |