The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002230.2 | NP_002221.1 | 2 - 3 | 341 | 78 |
NM_021991.2 | NP_068831.1 | 2 - 3 | 341 | 78 | |
XM_006721873.2 | XP_006721936.1 | 3 - 4 | 591 | 78 | |
XM_006721874.2 | XP_006721937.1 | 2 - 3 | 504 | 78 | |
XM_006721875.1 | XP_006721938.1 | 2 - 3 | 423 | 78 | |
XM_006721878.1 | XP_006721941.1 | 2 - 3 | 404 | 78 | |
XM_011524753.2 | XP_011523055.1 | 3 - 4 | 627 | 78 | |
XM_011524755.1 | XP_011523057.1 | 2 - 3 | 336 | 78 | |
XM_011524756.1 | XP_011523058.1 | 2 - 3 | 234 | 78 | |
XM_011524757.2 | XP_011523059.1 | 2 - 3 | 501 | 78 | |
XM_011524758.1 | XP_011523060.1 | 2 - 3 | 230 | 78 | |
XM_017024588.1 | XP_016880077.1 | 2 - 3 | 323 | 78 | |
XM_017024589.1 | XP_016880078.1 | 2 - 3 | 324 | 78 | |
XM_017024590.1 | XP_016880079.1 | 2 - 3 | 230 | 78 | |
GenBank mRNA | AK226051.1 | - | 2 - 3 | 318 | 78 |
KU178155.1 | - | 1 - 2 | 208 | 78 |