The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010919.2 | NP_001010919.1 | 2 - 3 | 638 | 72 |
XM_005266999.3 | XP_005267056.1 | 1 - 2 | 922 | 72 | |
GenBank mRNA | BC146842.1 | - | 1 - 2 | 564 | 72 |
DN997293.1 | - | 1 - 2 | 554 | 72 |