The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098478.1 | NP_001091948.1 | 2 - 2 | 442 | 69 |
NM_001098479.1 | NP_001091949.1 | 2 - 2 | 442 | 69 | |
NM_018950.2 | NP_061823.2 | 2 - 2 | 442 | 69 | |
XM_011514564.1 | XP_011512866.1 | 2 - 2 | 348 | 69 | |
XM_017010810.1 | XP_016866299.1 | 2 - 2 | 348 | 69 | |
XM_017010811.1 | XP_016866300.1 | 2 - 2 | 348 | 69 | |
XM_017010812.1 | XP_016866301.1 | 2 - 2 | 348 | 69 | |
XM_017010813.1 | XP_016866302.1 | 2 - 2 | 348 | 69 | |
GenBank mRNA | AK096962.1 | - | 2 - 2 | 348 | 69 |
AK225185.1 | - | 2 - 2 | 346 | 69 | |
AK298704.1 | - | 2 - 2 | 348 | 69 | |
AK300090.1 | - | 2 - 2 | 381 | 69 | |
AK302420.1 | - | 3 - 3 | 312 | 69 | |
AK309156.1 | - | 2 - 2 | 348 | 69 | |
AY216682.1 | - | 2 - 2 | 306 | 69 | |
AY221102.1 | - | 2 - 2 | 306 | 69 | |
AY253269.1 | - | 2 - 2 | 318 | 69 | |
AY253270.1 | - | 2 - 2 | 318 | 69 | |
AY253271.1 | - | 2 - 2 | 318 | 69 | |
BC009260.2 | - | 2 - 2 | 374 | 69 | |
BC062991.1 | - | 2 - 2 | 322 | 69 | |
CT002787.1 | - | 2 - 2 | 361 | 69 | |
DQ367723.1 | - | 2 - 2 | 318 | 69 | |
GQ891552.1 | - | 2 - 2 | 318 | 69 |