Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIRLET7F1
- Entrez Gene ID
-
406888
- Gene Name
- microRNA let-7f-1
- Gene Aliases
-
LET7F1, MIRNLET7F1, let-7f-1
- Chromosome Location
-
Chr.9: 94176347 - 94176433 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029483.1 |
|
1 - 1 |
48 |
86 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA let-7a-1 |
MIRLET7A1 |
LET7A1, MIRNLET7A1, let-7a-1 |
-311.0
|
Sense |
Chr. - 94175957 - 94176036 on Build GRCh38 |
Downstream |
MIRLET7D host gene |
MIRLET7DHG |
|
136.0
|
Anti-sense |
Chr. - 94176569 - 94204566 on Build GRCh38 |
View Full Product Details