Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TMED11P
- Entrez Gene ID
-
100379220
- Gene Name
- transmembrane p24 trafficking protein 11, pseudogene
- Gene Aliases
-
p24a1, p24alpha1
- Chromosome Location
-
Chr.4: 1115197 - 1123164 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033768.1 |
|
3 - 4 |
314 |
143 |
GenBank mRNA |
BG190838.1 |
- |
1 - 2 |
147 |
143 |
BG215779.1 |
- |
2 - 3 |
312 |
143 |
DY654917.1 |
- |
1 - 2 |
148 |
143 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105374344 |
LOC105374344 |
|
|
Anti-sense |
Chr. - 1113639 - 1132975 on Build GRCh38 |
Downstream |
spondin 2 |
SPON2 |
DIL-1, DIL1, M-SPONDIN, MINDIN |
50.5kb
|
Sense |
Chr. - 1166932 - 1208962 on Build GRCh38 |
View Full Product Details