Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIST2H2BC
Entrez Gene ID
337873
Gene Name
histone cluster 2, H2bc (pseudogene)
Gene Aliases
H2B/t, HIST2H2BD
Chromosome Location
Chr.1: 149850192 - 149850773 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_036461.1 1 - 1 519 102
GenBank mRNA AK299891.1 - 1 - 1 683 102


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY074794.1 - 1 245 Chr. 1 - 149850001 - 149850496 [-] on Build GRCh38 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream histone cluster 2, H2bd (pseudogene) HIST2H2BD H2B/o, H2B/s, H2BFO -6.7kb Anti-sense Chr. - 149843039 - 149843510 on Build GRCh38
Downstream histone cluster 2, H2aa4 HIST2H2AA4 H2A/R 288.0 Anti-sense Chr. - 149851061 - 149851594 on Build GRCh38

View Full Product Details