Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZBTB20-AS1
- Entrez Gene ID
-
100131117
- Gene Name
- ZBTB20 antisense RNA 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 114351811 - 114388978 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_038993.1 |
|
4 - 5 |
495 |
77 |
GenBank mRNA |
AK098660.1 |
- |
4 - 5 |
471 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 568 |
MIR568 |
MIRN568, hsa-mir-568 |
-42.8kb
|
Anti-sense |
Chr. - 114316475 - 114316569 on Build GRCh38 |
Downstream |
uncharacterized LOC101929754 |
LOC101929754 |
|
86.7kb
|
Sense |
Chr. - 114453593 - 114520132 on Build GRCh38 |
View Full Product Details