Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2G1P
Entrez Gene ID
22952
Gene Name
cytochrome P450 family 2 subfamily G member 1, pseudogene
Gene Aliases
CYP2G1, CYP2G2P, CYP2GP1
Chromosome Location
Chr.19: 40890826 - 40900508 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_040249.1 2 - 3 1647 78
GenBank mRNA AK127151.1 - 2 - 3 78


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN266537.1 - 3 - 4 Chr. 19 - 40801297 - 40898282 [+] on Build GRCh38 78
GenBankmRNA DR008188.1 - 3 - 4 Chr. 19 - 40893947 - 40900022 [-] on Build GRCh38 78


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream cytochrome P450 family 2 subfamily A member 7 CYP2A7 CPA7, CPAD, CYP2A, CYPIIA7, P450-IIA4 -10.9kb Anti-sense Chr. - 40875439 - 40885952 on Build GRCh38
Downstream cytochrome P450 family 2 subfamily B member 7, pseudogene CYP2B7P CYP2B, CYP2B7, CYP2B7P1 25.9kb Sense Chr. - 40924265 - 40950660 on Build GRCh38

View Full Product Details