Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BOK-AS1
- Entrez Gene ID
-
100379249
- Gene Name
- BOK antisense RNA 1
- Gene Aliases
-
BOK-AS, BOKAS, NAToB, NCRNA00151
- Chromosome Location
-
Chr.2: 241544384 - 241559143 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033346.1 |
|
1 - 1 |
126 |
111 |
GenBank mRNA |
FJ185168.1 |
- |
1 - 1 |
126 |
111 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AL565705.3 |
- |
1 |
|
|
Chr. 2 - 241558600 - 241559454 [+] on Build GRCh38 |
111 |
GenBankmRNA |
CR619343.1 |
- |
1 |
656 |
|
Chr. 2 - 241558453 - 241559673 [-] on Build GRCh38 |
111 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
serine/threonine kinase 25 |
STK25 |
SOK1, YSK1 |
-49.0kb
|
Sense |
Chr. - 241494707 - 241509730 on Build GRCh38 |
Downstream |
THAP domain containing 4 |
THAP4 |
CGI-36, PP238 |
25.3kb
|
Sense |
Chr. - 241584405 - 241637543 on Build GRCh38 |
View Full Product Details