The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244664.1 | NP_001231593.1 | 3 - 4 | 597 | 80 |
GenBank mRNA | AK095230.1 | - | 2 - 3 | 2145 | 80 |
BP375397.1 | - | 3 - 4 | 504 | 80 |