The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244847.1 | NP_001231776.1 | 2 - 3 | 214 | 79 |
GenBank mRNA | W95920.1 | - | 2 - 3 | 123 | 79 |