The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003817.3 | NP_003808.2 | 8 - 9 | 812 | 188 |
XM_017013943.1 | XP_016869432.1 | 8 - 9 | 699 | 188 | |
GenBank mRNA | AF090327.1 | - | 3 - 4 | 201 | 188 |
AF215824.1 | - | 8 - 9 | 783 | 188 | |
AK292492.1 | - | 8 - 9 | 816 | 188 | |
BC043207.2 | - | 2 - 3 | 145 | 188 |