The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003817.3 | NP_003808.2 | 9 - 10 | 986 | 122 |
XM_017013943.1 | XP_016869432.1 | 9 - 10 | 873 | 122 | |
GenBank mRNA | AF090327.1 | - | 4 - 5 | 375 | 122 |
AF215824.1 | - | 9 - 10 | 957 | 122 | |
AK292492.1 | - | 9 - 10 | 990 | 122 | |
BC043207.2 | - | 3 - 4 | 319 | 122 |