Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF3
Entrez Gene ID
22806
Gene Name
IKAROS family zinc finger 3
Gene Aliases
AIO, AIOLOS, ZNFN1A3
Chromosome Location
Chr.17: 39757715 - 39864188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257410.1 NP_001244339.1 3 - 4 226 66
GenBank mRNA AY377976.1 - 3 - 4 226 66

View Full Product Details