Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
Chromosome Location
Chr.7: 81699006 - 81770438 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000601.5 NP_000592.3 3 - 3 577 90
NM_001010931.2 NP_001010931.1 3 - 3 577 90
NM_001010932.2 NP_001010932.1 3 - 3 577 90
NM_001010933.2 NP_001010933.1 3 - 3 577 90
NM_001010934.2 NP_001010934.1 3 - 3 577 90
XM_006715956.2 XP_006716019.1 4 - 4 540 90
XM_011516115.2 XP_011514417.1 4 - 4 540 90
XM_017012097.1 XP_016867586.1 4 - 4 540 90
XM_017012098.1 XP_016867587.1 4 - 4 540 90
GenBank mRNA AB208900.1 - 3 - 3 484 90
AK291674.1 - 3 - 3 512 90
AK313694.1 - 3 - 3 427 90
BC022308.1 - 3 - 3 426 90
BC063485.1 - 3 - 3 435 90
BC105797.1 - 3 - 3 517 90
BC130284.1 - 3 - 3 425 90
BC130286.1 - 3 - 3 425 90
FJ830861.1 - 3 - 3 350 90
FJ830862.1 - 3 - 3 350 90
L02931.1 - 3 - 3 410 90
M29145.1 - 3 - 3 451 90
M55379.1 - 3 - 3 350 90
M60718.1 - 3 - 3 422 90
M73239.1 - 3 - 3 415 90
M73240.1 - 3 - 3 415 90
M77227.1 - 3 - 3 432 90
U46010.1 - 3 - 3 404 90
X57574.1 - 3 - 3 427 90

View Full Product Details