Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CELF2-AS2
- Entrez Gene ID
-
439950
- Gene Name
- CELF2 antisense RNA 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 11071541 - 11105488 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_046108.1 |
|
2 - 3 |
424 |
106 |
GenBank mRNA |
BC046381.1 |
- |
2 - 3 |
426 |
106 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB061959.1 |
- |
2 - 3 |
429 |
|
Chr. 10 - 11097285 - 11105494 [-] on Build GRCh38 |
106 |
GenBankmRNA |
BG723461.1 |
- |
|
431 |
|
Chr. - - [-] on Build |
106 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 710 |
LINC00710 |
|
-145.1kb
|
Sense |
Chr. - 10934940 - 10952163 on Build GRCh38 |
Downstream |
CELF2 antisense RNA 1 |
CELF2-AS1 |
C10orf31 |
218.4kb
|
Sense |
Chr. - 11316833 - 11344674 on Build GRCh38 |
View Full Product Details