The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_046512.2 | 1 - 2 | 93 | 79 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | SAM domain, SH3 domain and nuclear localization signals 1 | SAMSN1 | HACS1, NASH1, SASH2, SH3D6B, SLy2 | Anti-sense | Chr. - 14485228 - 14583402 on Build GRCh38 | |
Downstream | uncharacterized protein ENSP00000383407-like | LOC388813 | 5.9kb | Anti-sense | Chr. - 14591392 - 14643107 on Build GRCh38 |