Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC9A9-AS1
- Entrez Gene ID
-
100885796
- Gene Name
- SLC9A9 antisense RNA 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 143342246 - 143347071 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_048544.1 |
|
2 - 3 |
304 |
111 |
GenBank mRNA |
DB338547.1 |
- |
2 - 3 |
|
111 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 9 member A9 |
SLC9A9 |
AUTS16, NHE9 |
|
Anti-sense |
Chr. - 143265222 - 143848531 on Build GRCh38 |
Downstream |
chromosome 3 open reading frame 58 |
C3orf58 |
DIA1, GoPro49, HASF |
627.2kb
|
Sense |
Chr. - 143971798 - 143992368 on Build GRCh38 |
View Full Product Details