Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A14
Entrez Gene ID
84689
Gene Name
membrane spanning 4-domains A14
Gene Aliases
MS4A16, NYD-SP21
Chromosome Location
Chr.11: 60396014 - 60417756 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_049731.1 3 - 4 979 84
NR_049732.1 4 - 5 1030 84
NR_049733.1 3 - 4 979 84
NR_049734.1 4 - 5 1030 84
GenBank mRNA AK057418.1 - 3 - 4 525 84
BC107431.1 - 4 - 5 535 84
DB450359.2 - 4 - 5 538 84


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ435535.1 - 4 - 5 534 Chr. 11 - 60396508 - 60415499 [+] on Build GRCh38 84
GenBankmRNA BI560622.1 - 3 - 4 503 Chr. 11 - 60396494 - 60415487 [+] on Build GRCh38 84
GenBankmRNA BM554322.1 - 4 - 5 562 Chr. 11 - 60396480 - 60415745 [+] on Build GRCh38 84
GenBankmRNA DB454578.2 - 4 - 5 559 Chr. 11 - 60396485 - 60405933 [+] on Build GRCh38 84
GenBankmRNA BI560592.1 - 4 - 5 562 Chr. 11 - 60396487 - 60415605 [+] on Build GRCh38 84
GenBankmRNA BI825801.1 - 3 - 4 510 Chr. 11 - 60396481 - 60415501 [+] on Build GRCh38 84
GenBankmRNA BI562282.1 - 4 - 5 562 Chr. 11 - 60396486 - 60405915 [+] on Build GRCh38 83


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream membrane spanning 4-domains A7 MS4A7 4SPAN2, CD20L4, CFFM4, MS4A8 -7.0kb Sense Chr. - 60378485 - 60395954 on Build GRCh38
Downstream membrane spanning 4-domains A5 MS4A5 CD20-L2, CD20L2, TETM4 24.9kb Sense Chr. - 60429589 - 60447792 on Build GRCh38

View Full Product Details