Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX17
Entrez Gene ID
9784
Gene Name
sorting nexin 17
Gene Aliases
-
Chromosome Location
Chr.2: 27370496 - 27377533 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267059.1 NP_001253988.1 4 - 5 529 85
NM_001267060.1 NP_001253989.1 3 - 4 490 85
NM_001267061.1 NP_001253990.1 4 - 5 400 85
NM_014748.3 NP_055563.1 4 - 5 565 85
NR_049782.1 4 - 5 690 85
NR_049783.1 5 - 6 663 85
NR_049784.1 5 - 6 639 85
NR_049785.1 3 - 4 572 85
NR_049786.1 4 - 5 521 85
NR_049787.1 2 - 3 372 85
NR_049788.1 2 - 3 372 85
XM_011533203.1 XP_011531505.1 3 - 4 339 85
XM_017005405.1 XP_016860894.1 3 - 4 339 85
GenBank mRNA AK129689.1 - 4 - 5 449 85
AK222543.1 - 4 - 5 448 85
AK293252.1 - 5 - 6 539 85
AK293464.1 - 4 - 5 381 85
AK295278.1 - 2 - 3 251 85
AK297054.1 - 4 - 5 574 85
AK298620.1 - 4 - 5 400 85
AK298856.1 - 4 - 5 402 85
AK298869.1 - 3 - 4 351 85
AK300144.1 - 4 - 5 412 85
AK300641.1 - 2 - 3 245 85
AK308487.1 - 3 - 4 451 85
AK309640.1 - 3 - 4 323 85
AK316373.1 - 4 - 5 344 85
BC002524.1 - 4 - 5 426 85
BC002610.1 - 4 - 5 415 85
BC014620.2 - 4 - 5 426 85
BC050590.1 - 4 - 5 420 85
BT007167.1 - 4 - 5 317 85
CR457081.1 - 4 - 5 317 85
D31764.1 - 4 - 5 539 85

View Full Product Details