The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270707.1 | NP_001257636.1 | 1 - 2 | 287 | 69 |
NM_001270708.1 | NP_001257637.1 | 1 - 2 | 287 | 69 | |
NM_001270709.1 | NP_001257638.1 | 1 - 2 | 287 | 69 | |
NM_007243.2 | NP_009174.1 | 2 - 3 | 484 | 69 | |
NR_073065.1 | 1 - 2 | 287 | 69 | ||
GenBank mRNA | AF143676.1 | - | 1 - 2 | 130 | 69 |
AK075509.1 | - | 2 - 3 | 486 | 69 | |
AK315190.1 | - | 1 - 2 | 142 | 69 | |
AY358411.1 | - | 1 - 2 | 151 | 69 | |
BC021260.1 | - | 1 - 2 | 122 | 69 | |
BC039865.1 | - | 1 - 2 | 150 | 69 | |
BM847576.1 | - | 1 - 2 | 161 | 69 | |
BM917173.1 | - | 1 - 2 | 144 | 69 | |
BU159435.1 | - | 1 - 2 | 96 | 69 | |
JQ798186.1 | - | 1 - 2 | 150 | 69 |