Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MEAF6
- Entrez Gene ID
-
64769
- Gene Name
- MYST/Esa1 associated factor 6
- Gene Aliases
-
C1orf149, CENP-28, EAF6, NY-SAR-91
- Chromosome Location
-
Chr.1: 37489960 - 37514819 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073092.1 |
|
4 - 5 |
412 |
155 |
GenBank mRNA |
AK309663.1 |
- |
4 - 5 |
373 |
155 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA692707.1 |
- |
4 - 5 |
373 |
|
Chr. 1 - 37501883 - 37514780 [-] on Build GRCh38 |
155 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 5581 |
MIR5581 |
|
-4.1kb
|
Sense |
Chr. - 37500935 - 37500994 on Build GRCh38 |
Downstream |
Smad nuclear interacting protein 1 |
SNIP1 |
PMRED |
25.1kb
|
Sense |
Chr. - 37534449 - 37554344 on Build GRCh38 |
View Full Product Details