Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NHLRC3
- Entrez Gene ID
-
387921
- Gene Name
- NHL repeat containing 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 39038311 - 39050109 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073109.1 |
|
1 - 2 |
157 |
70 |
GenBank mRNA |
BC052492.1 |
- |
1 - 2 |
95 |
70 |
DA772703.1 |
- |
1 - 2 |
157 |
70 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG719308.1 |
- |
1 - 2 |
100 |
|
Chr. 13 - 39038373 - 39044144 [+] on Build GRCh38 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
proline and serine rich 1 |
PROSER1 |
C13orf23 |
-216.0
|
Anti-sense |
Chr. - 39009865 - 39038115 on Build GRCh38 |
Downstream |
lipoma HMGIC fusion partner |
LHFP |
|
303.6kb
|
Anti-sense |
Chr. - 39342892 - 39603219 on Build GRCh38 |
View Full Product Details