Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EPGN
- Entrez Gene ID
-
255324
- Gene Name
- epithelial mitogen
- Gene Aliases
-
ALGV3072, EPG, PRO9904
- Chromosome Location
-
Chr.4: 74307571 - 74317298 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073117.1 |
|
1 - 2 |
109 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105377276 |
LOC105377276 |
|
|
Anti-sense |
Chr. - 74278045 - 74330322 on Build GRCh38 |
Downstream |
epiregulin |
EREG |
EPR, ER, Ep |
52.8kb
|
Sense |
Chr. - 74365143 - 74388760 on Build GRCh38 |
View Full Product Details