Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2W
Entrez Gene ID
55284
Gene Name
ubiquitin conjugating enzyme E2 W (putative)
Gene Aliases
UBC-16, UBC16
Chromosome Location
Chr.8: 73780097 - 73878910 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_073119.1 5 - 6 554 190
GenBank mRNA DA352184.1 - 5 - 6 507 190


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream staufen double-stranded RNA binding protein 2 STAU2 39K2, 39K3 -38.5kb Sense Chr. - 73420369 - 73747708 on Build GRCh38
Downstream long intergenic non-protein coding RNA 1617 LINC01617 C8orf23, NCRNA00249, XLOC_006844 83.1kb Anti-sense Chr. - 73879010 - 73904730 on Build GRCh38

View Full Product Details