Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CDH19
- Entrez Gene ID
-
28513
- Gene Name
- cadherin 19
- Gene Aliases
-
CDH7, CDH7L2
- Chromosome Location
-
Chr.18: 66501187 - 66604241 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073130.1 |
|
10 - 11 |
1878 |
150 |
GenBank mRNA |
AK315257.1 |
- |
10 - 11 |
1880 |
150 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cadherin 7 |
CDH7 |
CDH7L1 |
-628.1kb
|
Anti-sense |
Chr. - 65750252 - 65885136 on Build GRCh38 |
Downstream |
microRNA 5011 |
MIR5011 |
|
569.9kb
|
Anti-sense |
Chr. - 67081584 - 67081686 on Build GRCh38 |
View Full Product Details