Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PGR
- Entrez Gene ID
-
5241
- Gene Name
- progesterone receptor
- Gene Aliases
-
NR3C3, PR
- Chromosome Location
-
Chr.11: 101029624 - 101130681 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073142.2 |
|
2 - 3 |
1781 |
95 |
GenBank mRNA |
AB085843.1 |
- |
2 - 3 |
1788 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
transmembrane protein 133 |
TMEM133 |
AD031 |
-68.5kb
|
Anti-sense |
Chr. - 100992080 - 100993935 on Build GRCh38 |
Downstream |
progesterone receptor antisense RNA |
LOC101054525 |
|
2.9kb
|
Anti-sense |
Chr. - 101129077 - 101159270 on Build GRCh38 |
View Full Product Details