Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FAM90A2P
- Entrez Gene ID
-
729689
- Gene Name
- family with sequence similarity 90, member A2 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 12173454 - 12177267 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_046354.1 |
|
1 - 2 |
35 |
72 |
GenBank mRNA |
CX762572.1 |
- |
1 - 2 |
71 |
72 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
family with sequence similarity 66 member D |
FAM66D |
|
-24.5kb
|
Anti-sense |
Chr. - 12115782 - 12151189 on Build GRCh38 |
Downstream |
family with sequence similarity 86 member B1 |
FAM86B1 |
|
4.8kb
|
Sense |
Chr. - 12182104 - 12194118 on Build GRCh38 |
View Full Product Details