Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTGS1
Entrez Gene ID
5742
Gene Name
prostaglandin-endoperoxide synthase 1
Gene Aliases
COX1, COX3, PCOX1, PES-1, PGG/HS, PGHS-1, PGHS1, PHS1, PTGHS
Chromosome Location
Chr.9: 122369906 - 122395703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000962.3 NP_000953.2 6 - 7 762 71
NM_001271164.1 NP_001258093.1 5 - 6 618 71
NM_001271165.1 NP_001258094.1 5 - 6 711 71
NM_001271166.1 NP_001258095.1 6 - 7 988 71
NM_001271367.1 NP_001258296.1 6 - 7 733 71
NM_001271368.1 NP_001258297.1 7 - 8 1210 71
NM_080591.2 NP_542158.1 6 - 7 762 71
XM_005252105.3 XP_005252162.1 7 - 8 937 71
XM_011518875.2 XP_011517177.1 7 - 8 1198 71
XM_011518876.2 XP_011517178.1 7 - 8 4808 71
XM_017014929.1 XP_016870418.1 9 - 10 1172 71
GenBank mRNA AK290022.1 - 6 - 7 715 71
AK295221.1 - 7 - 8 1195 71
AK304403.1 - 5 - 6 711 71
AK308222.1 - 6 - 7 988 71
BC029840.1 - 6 - 7 734 71
CD014048.1 - 5 - 6 584 71
CD014049.1 - 6 - 7 699 71
DQ180741.1 - 5 - 6 775 71
DQ180742.1 - 5 - 6 775 71
M59979.1 - 5 - 6 687 71
S36219.1 - 5 - 6 682 71
S36271.1 - 5 - 6 682 71
S78220.1 - 5 - 6 682 71

View Full Product Details