Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OBSCN
Entrez Gene ID
84033
Gene Name
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases
ARHGEF30, UNC89
Chromosome Location
Chr.1: 228208013 - 228381431 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098623.2 NP_001092093.2 44 - 45 11997 87
NM_001271223.2 NP_001258152.2 55 - 56 14868 87
NM_052843.3 NP_443075.3 44 - 45 11997 87
XM_005273287.4 XP_005273344.1 54 - 55 14672 87
XM_005273291.4 XP_005273348.1 54 - 55 14672 87
XM_005273298.4 XP_005273355.1 54 - 55 14672 87
XM_005273307.4 XP_005273364.1 54 - 55 14672 87
XM_006711822.3 XP_006711885.1 54 - 55 14672 87
XM_006711823.3 XP_006711886.1 54 - 55 14672 87
XM_006711827.3 XP_006711890.1 48 - 49 13087 87
XM_006711829.3 XP_006711892.1 47 - 48 12811 87
XM_011544297.2 XP_011542599.1 48 - 49 13088 87
XM_011544299.2 XP_011542601.1 47 - 48 12812 87
XM_017002443.1 XP_016857932.1 54 - 55 14672 87
XM_017002444.1 XP_016857933.1 54 - 55 14635 87
XM_017002445.1 XP_016857934.1 54 - 55 14672 87
XM_017002446.1 XP_016857935.1 54 - 55 14672 87
XM_017002447.1 XP_016857936.1 54 - 55 14672 87
XM_017002448.1 XP_016857937.1 54 - 55 14522 87
XM_017002449.1 XP_016857938.1 53 - 54 14409 87
XM_017002450.1 XP_016857939.1 53 - 54 14409 87
XM_017002451.1 XP_016857940.1 53 - 54 14409 87
XM_017002452.1 XP_016857941.1 53 - 54 14409 87
XM_017002453.1 XP_016857942.1 53 - 54 14396 87
XM_017002454.1 XP_016857943.1 53 - 54 14396 87
XM_017002455.1 XP_016857944.1 53 - 54 14396 87
XM_017002456.1 XP_016857945.1 53 - 54 14396 87
XM_017002457.1 XP_016857946.1 53 - 54 14396 87
XM_017002458.1 XP_016857947.1 52 - 53 14145 87
XM_017002459.1 XP_016857948.1 52 - 53 14145 87
XM_017002460.1 XP_016857949.1 52 - 53 14145 87
XM_017002461.1 XP_016857950.1 54 - 55 14672 87
XM_017002462.1 XP_016857951.1 52 - 53 14120 87
XM_017002463.1 XP_016857952.1 52 - 53 14099 87
XM_017002464.1 XP_016857953.1 54 - 55 14672 87
XM_017002465.1 XP_016857954.1 51 - 52 13881 87
XM_017002466.1 XP_016857955.1 51 - 52 13881 87
XM_017002467.1 XP_016857956.1 51 - 52 13881 87
XM_017002468.1 XP_016857957.1 50 - 51 13617 87
XM_017002469.1 XP_016857958.1 48 - 49 13089 87
XM_017002470.1 XP_016857959.1 43 - 44 11770 87
XM_017002471.1 XP_016857960.1 54 - 55 14671 87
XM_017002472.1 XP_016857961.1 54 - 55 14671 87
XM_017002473.1 XP_016857962.1 54 - 55 14671 87
GenBank mRNA AJ002535.1 - 45 - 46 11994 87

View Full Product Details