Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OBSCN
Entrez Gene ID
84033
Gene Name
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases
ARHGEF30, UNC89
Chromosome Location
Chr.1: 228208013 - 228381431 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098623.2 NP_001092093.2 51 - 52 13639 60
NM_001271223.2 NP_001258152.2 62 - 63 16510 60
NM_052843.3 NP_443075.3 51 - 52 13639 60
XM_005273287.4 XP_005273344.1 63 - 64 16890 60
XM_005273291.4 XP_005273348.1 63 - 64 16854 60
XM_005273298.4 XP_005273355.1 62 - 63 16581 60
XM_005273307.4 XP_005273364.1 58 - 59 15768 60
XM_006711822.3 XP_006711885.1 61 - 62 16314 60
XM_006711823.3 XP_006711886.1 61 - 62 16314 60
XM_006711827.3 XP_006711890.1 55 - 56 14729 60
XM_006711829.3 XP_006711892.1 54 - 55 14453 60
XM_011544297.2 XP_011542599.1 55 - 56 14730 60
XM_011544299.2 XP_011542601.1 54 - 55 14454 60
XM_017002443.1 XP_016857932.1 63 - 64 16935 60
XM_017002444.1 XP_016857933.1 63 - 64 16898 60
XM_017002445.1 XP_016857934.1 63 - 64 16935 60
XM_017002446.1 XP_016857935.1 63 - 64 16935 60
XM_017002447.1 XP_016857936.1 63 - 64 16935 60
XM_017002448.1 XP_016857937.1 63 - 64 16785 60
XM_017002449.1 XP_016857938.1 62 - 63 16672 60
XM_017002450.1 XP_016857939.1 62 - 63 16672 60
XM_017002451.1 XP_016857940.1 62 - 63 16672 60
XM_017002452.1 XP_016857941.1 62 - 63 16672 60
XM_017002453.1 XP_016857942.1 62 - 63 16659 60
XM_017002454.1 XP_016857943.1 62 - 63 16659 60
XM_017002455.1 XP_016857944.1 62 - 63 16659 60
XM_017002456.1 XP_016857945.1 62 - 63 16659 60
XM_017002457.1 XP_016857946.1 62 - 63 16659 60
XM_017002458.1 XP_016857947.1 61 - 62 16408 60
XM_017002459.1 XP_016857948.1 61 - 62 16408 60
XM_017002460.1 XP_016857949.1 61 - 62 16408 60
XM_017002461.1 XP_016857950.1 61 - 62 16395 60
XM_017002462.1 XP_016857951.1 61 - 62 16383 60
XM_017002463.1 XP_016857952.1 61 - 62 16362 60
XM_017002464.1 XP_016857953.1 61 - 62 16314 60
XM_017002465.1 XP_016857954.1 60 - 61 16144 60
XM_017002466.1 XP_016857955.1 60 - 61 16144 60
XM_017002467.1 XP_016857956.1 60 - 61 16144 60
XM_017002468.1 XP_016857957.1 59 - 60 15880 60
XM_017002469.1 XP_016857958.1 57 - 58 15352 60
XM_017002470.1 XP_016857959.1 52 - 53 14033 60
XM_017002471.1 XP_016857960.1 63 - 64 16934 60
XM_017002472.1 XP_016857961.1 63 - 64 16934 60
XM_017002473.1 XP_016857962.1 63 - 64 16934 60

View Full Product Details