The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270989.1 | NP_001257918.1 | 4 - 5 | 477 | 104 |
XM_011531817.2 | XP_011530119.1 | 4 - 5 | 1335 | 104 | |
GenBank mRNA | AK289455.1 | - | 4 - 5 | 475 | 104 |
DQ235269.1 | - | 4 - 5 | 461 | 104 |