The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271006.1 | NP_001257935.1 | 2 - 3 | 138 | 71 |
XM_005256339.2 | XP_005256396.1 | 2 - 3 | 159 | 71 | |
XM_017024326.1 | XP_016879815.1 | 2 - 3 | 159 | 71 | |
XM_017024327.1 | XP_016879816.1 | 2 - 3 | 159 | 71 | |
XM_017024328.1 | XP_016879817.1 | 2 - 3 | 159 | 71 | |
GenBank mRNA | CB110145.1 | - | 2 - 3 | 147 | 71 |
HY110543.1 | - | 2 - 3 | 128 | 71 |