Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCGR3B
Entrez Gene ID
2215
Gene Name
Fc fragment of IgG receptor IIIb
Gene Aliases
CD16, CD16b, FCG3, FCGR3, FCR-10, FCRIII, FCRIIIb
Chromosome Location
Chr.1: 161623196 - 161631963 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271037.1 NP_001257966.1 3 - 4 554 64

View Full Product Details