Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- BCL2L13
- Entrez Gene ID
-
23786
- Gene Name
- BCL2 like 13
- Gene Aliases
-
BCL-RAMBO, Bcl2-L-13, MIL1
- Chromosome Location
-
Chr.22: 17628855 - 17730855 on Build GRCh38
View Full Product Details