The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271329.1 | NP_001258258.1 | 1 - 2 | 383 | 72 |
GenBank mRNA | AK307585.1 | - | 1 - 2 | 352 | 72 |