Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACBD5
Entrez Gene ID
91452
Gene Name
acyl-CoA binding domain containing 5
Gene Aliases
-
Chromosome Location
Chr.10: 27182624 - 27248674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042473.3 NP_001035938.1 8 - 9 1003 99
NM_001271512.3 NP_001258441.1 7 - 8 1317 99
NM_001301251.1 NP_001288180.1 7 - 8 882 99
NM_001301252.1 NP_001288181.1 6 - 7 1196 99
NM_001301253.1 NP_001288182.1 7 - 8 872 99
NM_001301254.1 NP_001288183.1 5 - 6 992 99
NM_145698.4 NP_663736.2 8 - 9 1128 99
XM_006717528.3 XP_006717591.3 9 - 10 7561 99
XM_006717530.3 XP_006717593.1 9 - 10 1160 99
XM_006717531.2 XP_006717594.1 9 - 10 1170 99
XM_006717535.2 XP_006717598.1 9 - 10 1027 99
XM_017016884.1 XP_016872373.1 9 - 10 7561 99
XM_017016885.1 XP_016872374.1 8 - 9 7528 99
XM_017016886.1 XP_016872375.1 8 - 9 7528 99
XM_017016887.1 XP_016872376.1 9 - 10 1091 99
XM_017016888.1 XP_016872377.1 9 - 10 1156 99
XM_017016889.1 XP_016872378.1 8 - 9 1058 99
XM_017016890.1 XP_016872379.1 8 - 9 7357 99
XM_017016891.1 XP_016872380.1 8 - 9 1123 99
XM_017016892.1 XP_016872381.1 8 - 9 1044 99
XM_017016893.1 XP_016872382.1 7 - 8 7324 99
XM_017016894.1 XP_016872383.1 7 - 8 7324 99
XM_017016895.1 XP_016872384.1 9 - 10 1027 99
XM_017016896.1 XP_016872385.1 8 - 9 995 99
XM_017016897.1 XP_016872386.1 8 - 9 952 99
XM_017016898.1 XP_016872387.1 7 - 8 919 99
XM_017016899.1 XP_016872388.1 7 - 8 919 99
XM_017016900.1 XP_016872389.1 8 - 9 945 99
XM_017016901.1 XP_016872390.1 8 - 9 925 99
XM_017016902.1 XP_016872391.1 8 - 9 944 99
XM_017016903.1 XP_016872392.1 8 - 9 929 99
XM_017016904.1 XP_016872393.1 7 - 8 924 99
XM_017016905.1 XP_016872394.1 7 - 8 910 99
XM_017016906.1 XP_016872395.1 7 - 8 791 99
XM_017016907.1 XP_016872396.1 7 - 8 924 99
XM_017016908.1 XP_016872397.1 7 - 8 913 99
GenBank mRNA AB082527.1 - 7 - 8 831 99
AF505653.1 - 7 - 8 869 99
AK057469.1 - 8 - 9 995 99
AK294510.1 - 5 - 6 910 99
AK295031.1 - 7 - 8 954 99
AK304842.1 - 6 - 7 847 99
AL133064.1 - 2 - 3 211 99
BC030555.1 - 8 - 9 1032 99

View Full Product Details